Learn More
Aviva Systems Biology Corp ZNF641 Antibody - middle region (ARP39880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF641. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF641. Alternate names: DKFZp667D1012, FLJ31295ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57215