Learn More
Aviva Systems Biology Corp ZNF644 Antibody - middle region (ARP39684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF644. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF644. Alternate names: BM-005, KIAA1221, MGC60165, MGC70410, Zep-2, NatF, MYP21, ZEP-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53150