Learn More
Aviva Systems Biology Corp CBLL2 Antibody - N-terminal region (ARP36191_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36191_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF645. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF645. Alternate names: HAKAILApplicationWB. Species reactivity: Human
Catalog No. 50-014-56059