Learn More
Aviva Systems Biology Corp ZNF646 Antibody - N-terminal region (ARP33712_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33712_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF646. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF646. Alternate names: KIAA0296ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-55123