Learn More
Aviva Systems Biology Corp ZNF648 Antibody - N-terminal region (ARP50356_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50356_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF648. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF648. Alternate names: -ApplicationWB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-014-60326