Learn More
Aviva Systems Biology Corp ZNF652 Antibody - N-terminal region (ARP31678_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31678_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF652. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF652. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-52835