Learn More
Aviva Systems Biology Corp ZNF654 Antibody - N-terminal region (ARP33145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF654. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF654. Alternate names: FLJ10997, FLJ21142ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-52895