Learn More
Aviva Systems Biology Corp ZNF654 Antibody - middle region (ARP33144_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33144_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF654. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF654. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-52894