Learn More
Aviva Systems Biology Corp ZNF655 Antibody - N-terminal region (ARP39587_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39587_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF655. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF655. Alternate names: VIK, VIK-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-57096