Learn More
Aviva Systems Biology Corp ZNF655 Antibody - middle region (ARP39818_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39818_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF655. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF655. Alternate names: DKFZp686M1631, FLJ23461, MGC10859, MGC16203, MGC5521, VIK, VIK-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57190