Learn More
Aviva Systems Biology Corp ZNF660 Antibody - C-terminal region (ARP36242_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36242_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF660. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF660. Alternate names: ApplicationIHC, WB. Species reactivity: Horse, Human, Rabbit, Rat
Catalog No. 50-014-56087