Learn More
Aviva Systems Biology Corp ZNF664 Antibody - N-terminal region (ARP51714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF664. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF664. Alternate names: DKFZp761B128, MGC126579, ZFOC1, ZNF176ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-60595