Learn More
Aviva Systems Biology Corp ZNF665 Antibody - N-terminal region (ARP36042_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36042_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF665. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF665. Alternate names: ZFP160LApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-55985