Learn More
Aviva Systems Biology Corp Znf667 Antibody - N-terminal region (ARP40087_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40087_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Znf667. It was validated on Western Blot. . Alternate names: Mip1, Zfp667, Znf667ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-852