Learn More
Aviva Systems Biology Corp ZNF667 Antibody - middle region (ARP36018_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36018_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF667. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF667. Alternate names: DKFZp686O111, FLJ14011, MIPU1ApplicationWB. Species reactivity: Cow, Horse, Human, Pig
Catalog No. 50-014-55972