Learn More
Aviva Systems Biology Corp ZNF668 Antibody - C-terminal region (ARP39611_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39611_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF668. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF668. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-57107