Learn More
Aviva Systems Biology Corp ZNF669 Antibody - C-terminal region (ARP39621_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39621_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF669. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF669. Alternate names: FLJ12606ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-57113