Learn More
Aviva Systems Biology Corp ZNF669 Antibody - N-terminal region (ARP36044_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36044_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF669. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF669. Alternate names: FLJ12606ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-55987