Learn More
Aviva Systems Biology Corp ZNF675 Antibody - C-terminal region (ARP36131_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36131_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF675. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF675. Alternate names: FLJ36350, TBZF, TIZApplicationWB. Species reactivity: Human
Catalog No. 50-014-53032