Learn More
Aviva Systems Biology Corp ZNF675 Antibody - N-terminal region (ARP57962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF675. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF675. Alternate names: FLJ36350, TBZF, TIZApplicationWB. Species reactivity: Human
Catalog No. 50-014-62194