Learn More
Aviva Systems Biology Corp ZNF677 Antibody - C-terminal region (ARP40061_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40061_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF677. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF677. Alternate names: MGC48625ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53171