Learn More
Aviva Systems Biology Corp ZNF677 Antibody - N-terminal region (ARP40060_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40060_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF677. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF677. Alternate names: MGC48625ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-284-849