Learn More
Aviva Systems Biology Corp ZNF678 Antibody - C-terminal region (ARP36255_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36255_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF678. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF678. Alternate names: FLJ18355, FLJ22822, MGC15634, MGC42493ApplicationWB. Species reactivity: Human
Catalog No. 50-284-488