Learn More
Aviva Systems Biology Corp ZNF681 Antibody - C-terminal region (ARP51701_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51701_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF681. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF681. Alternate names: FLJ31526ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53662