Learn More
Aviva Systems Biology Corp ZNF682 Antibody - N-terminal region (ARP39750_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39750_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF682. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF682. Alternate names: BC39498_3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57165