Learn More
Aviva Systems Biology Corp ZNF683 Antibody - N-terminal region (ARP36236_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36236_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF683. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF683. Alternate names: RP11-569G9.6ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-56082