Learn More
Aviva Systems Biology Corp ZNF683 Antibody - N-terminal region (ARP36237_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36237_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF683. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF683. Alternate names: MGC33414, RP11-569G9.6ApplicationWB. Species reactivity: Guinea Pig, Horse, Human
Catalog No. 50-014-56083