Learn More
Aviva Systems Biology Corp ZNF687 Antibody - C-terminal region (ARP39447_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39447_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF687. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF687. Alternate names: DKFZp781I1719, KIAA1441, RP11-126K1.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57033