Learn More
Aviva Systems Biology Corp ZNF688 Antibody - middle region (ARP36155_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36155_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF688. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZNF688. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-284-483