Learn More
Aviva Systems Biology Corp ZNF690 Antibody - middle region (ARP36179_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36179_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF690. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF690. Alternate names: ZNF690, Zfp690ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56051