Learn More
Aviva Systems Biology Corp ZNF691 Antibody - N-terminal region (ARP32076_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32076_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF691. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF691. Alternate names: Zfp691, RP11-342M1.5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54446