Learn More
Aviva Systems Biology Corp ZNF691 Antibody - N-terminal region (ARP39215_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39215_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF691. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF691. Alternate names: RP11-342M1.5, Zfp691ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-787