Learn More
Aviva Systems Biology Corp ZNF691 Antibody - middle region (ARP39216_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39216_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF691. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF691. Alternate names: RP11-342M1.5, Zfp691ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-56945