Learn More
Aviva Systems Biology Corp ZNF697 Antibody - N-terminal region (ARP51098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF697. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF697. Alternate names: MGC45731ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-556