Learn More
Aviva Systems Biology Corp ZNF701 Antibody - middle region (ARP35951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF701. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF701. Alternate names: FLJ10891ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-55941