Learn More
Aviva Systems Biology Corp ZNF703 Antibody - C-terminal region (ARP33640_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33640_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF703. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF703. Alternate names: FLJ14299, ZNF503L, ZPO1, ZEPPO1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55105