Learn More
Aviva Systems Biology Corp ZNF706 Antibody - N-terminal region (ARP32987_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32987_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF706. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF706. Alternate names: HSPC038, PNAS-106, PNAS-113ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54843