Learn More
Aviva Systems Biology Corp ZNF71 Antibody - middle region (ARP39498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF71. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF71. Alternate names: EZFITApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57057