Learn More
Aviva Systems Biology Corp ZNF711 Antibody - N-terminal region (ARP33508_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33508_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF711. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF711. Alternate names: CMPX1, ZNF4, ZNF5, ZNF6, Zfp711, dJ75N13.1, MRX97ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55047