Learn More
Aviva Systems Biology Corp ZNF746 Antibody - middle region (ARP32189_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32189_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF746. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF746. Alternate names: FLJ31413, PARISApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54479