Learn More
Aviva Systems Biology Corp ZNF76 Antibody - middle region (ARP33534_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33534_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF76. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF76. Alternate names: D6S229E, ZNF523, Zfp523ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55057