Learn More
Aviva Systems Biology Corp ZNF765 Antibody - N-terminal region (ARP58368_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58368_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF765. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF765. Alternate names: DKFZp686J2252ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-467