Learn More
Aviva Systems Biology Corp ZNF765 Antibody - middle region (ARP50446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF765. It was validated on Western Blot using a cell lysate as a positive control.. Alternate names: DKFZp686J2252ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-509