Learn More
Aviva Systems Biology Corp ZNF77 Antibody - N-terminal region (ARP36005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF77. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF77. Alternate names: pT1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55964