Learn More
Aviva Systems Biology Corp ZNF773 Antibody - N-terminal region (ARP34878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF773. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF773. Alternate names: FLJ00301, MGC4728, ZNF419BApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-55597