Learn More
Aviva Systems Biology Corp ZNF785 Antibody - C-terminal region (ARP36183_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36183_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF785. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF785. Alternate names: ApplicationIHC, WB. Species reactivity: Dog, Human
Catalog No. 50-014-56054