Learn More
Aviva Systems Biology Corp ZNF786 Antibody - N-terminal region (ARP39890_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39890_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF786. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF786. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53158