Learn More
Aviva Systems Biology Corp ZNF79 Antibody - N-terminal region (ARP35824_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35824_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF79. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF79. Alternate names: pT7ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55877