Learn More
Aviva Systems Biology Corp ZNF790 Antibody - N-terminal region (ARP50897_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50897_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF790. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF790. Alternate names: FLJ20350, MGC62100ApplicationWB. Species reactivity: Human
Catalog No. 50-285-549