Learn More
Aviva Systems Biology Corp ZNF791 Antibody - N-terminal region (ARP39954_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39954_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF791. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF791. Alternate names: FLJ90396, MGC126179, MGC126180ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-57244